Learn More
Perkin Elmer US LLC 1.5 mL Clear Glass 9 mm Screw Top High Recovery Vial, 100/pk
Catalog No. 502113229
Encompass_Preferred
Manufactured from the same high quality Type 1, Class A borosilicate glass as our standard 2 mL vials, these high recovery vials provide efficient handling with the convenience of a single vial format.
Catalog No. 50-211-3229 Supplier Perkin Elmer US LLC Supplier No. N2926202
Manufactured from the same high quality Type 1, Class A borosilicate glass as our standard 2 mL vials, these high recovery vials provide efficient handling with the convenience of a single vial format.