Learn More
Perkin Elmer US LLC 15 mL Graduated Conical Bottom SuperClear Tubes with Teal Flat Caps Included, Qty. 500
Catalog No. 502112193
Encompass_Preferred
Type: SuperClear Cap Type: Teal Flat Caps, Included Size: 17 mm x 118 mm Quantity: 500 (Bulk)
Catalog No. 50-211-2193 Supplier Perkin Elmer US LLC Supplier No. N0777703
Type: SuperClear Cap Type: Teal Flat Caps, Included Size: 17 mm x 118 mm Quantity: 500 (Bulk)