Learn More
Savillex Corporation Savillex 15 mL Standard Vial, High Purity Virgin PFA, Rounded Interior, Flat Bottom, 15 mL
SDP
Catalog No. NC1666896
Encompass
Savillex 15 mL standard vials offer exceptional chemical inertness and ultra-low trace metal content, ensuring sample integrity for critical scientific applications. Crafted from high-purity virgin PFA, these vials enhance laboratory efficiency with their rounded interior for complete contents removal and flat exterior bottom for efficient heating. Ideal for demanding trace metal analysis and sample storage, they stand out for their unmatched purity and performance in maintaining sample quality.
Catalog No. NC1666896 Supplier Savillex Corporation Supplier No. 20001520
Savillex 15 mL standard vials offer exceptional chemical inertness and ultra-low trace metal content, ensuring sample integrity for critical scientific applications. Crafted from high-purity virgin PFA, these vials enhance laboratory efficiency with their rounded interior for complete contents removal and flat exterior bottom for efficient heating. Ideal for demanding trace metal analysis and sample storage, they stand out for their unmatched purity and performance in maintaining sample quality.