Learn More
Electron Microscopy Sciences Syringe With Tips 15 ml, Autoclavable
SDP
Catalog No. 50949447
Encompass_Preferred
Used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. The 15 ml syringe dispenses 250 to 1250 uL.
Type: Autoclavable
Catalog No. 50-949-447 Supplier Electron Microscopy Sciences Supplier No. 72485
Used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. The 15 ml syringe dispenses 250 to 1250 uL.
Type: Autoclavable