Learn More
Labnet International, Inc 15 ML TUBE ROTISSERIE
Catalog No. NC9037020
Encompass_Preferred
Rotisserie for 10x15ml conical tubes or (15-16 mm dia)and 12x12-13mm dia tubes
Catalog No. NC9037020 Supplier Labnet International, Inc Supplier No. H550015
Rotisserie for 10x15ml conical tubes or (15-16 mm dia)and 12x12-13mm dia tubes