Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LifeSecure™ 15-Person 3-Day Infection Protection Kit
SDP

Catalog No. 19444609 Shop All Life Secure Products
Change view
Click to view available options
Color:
White, Blue, Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-444-609 White, Blue, Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-444-609 Supplier LifeSecure™ Supplier No. 12100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Infection protection supplies for 15 people can last for 3 days in case of pandemic or risk from infectious disease.

  • Protected in a tamper-evident stackable storage bucket
  • Includes N95 respirator masks, eye shields, vinyl gloves, hand sanitizer, tissue packs, biohazard bags, surface sanitizer spray, and disposable thermometers

Specifications

Product Type Infection Protection Kit
Color White, Blue, Orange
Material Plastic
Dimensions (L x W x H) 13 x 13.5 x 15 in.
Height (Metric) 18.1 cm
Length (Metric) 33.02 cm
Weight (English) 11 lb.
Weight (Metric) 4.99 kg
Width (Metric) 34.29 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.