Learn More
JDM LAB SOLUTIONS LLC 15 POSITION-MULTI MAG STIRRER
Catalog No. NC1866747
Encompass
15 Position Magnetic Stirrer. The front control panel has the ability to detach from the main unit. This unique feature allows the main stirring base to be placed & operated in a water bath.
Catalog No. NC1866747 Supplier JDM LAB SOLUTIONS LLC Supplier No. MS15BSUA
15 Position Magnetic Stirrer. The front control panel has the ability to detach from the main unit. This unique feature allows the main stirring base to be placed & operated in a water bath.