Learn More
Cole-Parmer Thermo Scientific Cimarec 50094596 i Poly 15 stirrer 100-240V
Catalog No. NC9032137 Shop All Cole-Parmer Products
Encompass
This stirrer is equipped with an electromagnetic drive, which means there is no moving parts and maintenance-free. It features a memory function that stores last speed settings.
Catalog No. NC9032137 Supplier Cole-Parmer Supplier No. 0465640
This stirrer is equipped with an electromagnetic drive, which means there is no moving parts and maintenance-free. It features a memory function that stores last speed settings.