Learn More
Cayman Chemical 15 R-LIpoxIn A4 100UG
SDP
Catalog No. 502894864
Encompass_Preferred
An aspirin-triggered lipoxin that inhibits LTB4-induced chemotaxis, adherence, and transmigration of the neutrophils with twice the potency of LXA4 demonstrating activity in the nM range
Catalog No. 50-289-4864 Supplier Cayman Chemical Supplier No. 90415100 UG
An aspirin-triggered lipoxin that inhibits LTB4-induced chemotaxis, adherence, and transmigration of the neutrophils with twice the potency of LXA4 demonstrating activity in the nM range