Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 15(R)-Lipoxin A4 | 171030-11-8 | 98.3% | 352.47 | 100 UG
SDP

Catalog No. 5000436667
Encompass_Preferred

15(R)-Lipoxin A4 | 171030-11-8 | 98.3% | 352.47 | 100 UG

Catalog No. 50-004-36667 Supplier Medchemexpress LLC Supplier No. HY113509B100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

15(R)-Lipoxin A4 | 171030-11-8 | 98.3% | 352.47 | 100 UG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.