Learn More
Cayman Chemical 15(S)-Fluprostenol, Purity: ≥98%, CAS: 54276-24-3, 10 mg
SDP
Catalog No. 502980064
Encompass_Preferred
An isomer of fluprostenol; a potential active metabolite to 15(S)-fluprostenol isopropyl ester
Catalog No. 50-298-0064 Supplier Cayman Chemical Supplier No. 1678710 MG
An isomer of fluprostenol; a potential active metabolite to 15(S)-fluprostenol isopropyl ester