Learn More
Enzo Life Sciences 15(S)-HETE ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900051
Encompass_Preferred
The 15(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 405 nm. Alternative Name: 15(S)-hydroperoxy tetraenoic eicosatetraenoic acid. Sensitivity: 69.21 pg/ml (range 78.1 - 20,000 pg/ml). Assay Time: Overnight + 1 hour. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-119