Learn More
Enzo Life Sciences 15(S)-HETE ELISA kit (5x96wells)
SDP
Catalog No. 502006890
Encompass_Preferred
The 15(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 405 nm. Alternative name: 15(S)-hydroperoxy tetraenoic eicosatetraenoic acid. Assay Time: Overnight + 1 hour. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
Catalog No. 50-200-6890 Supplier Enzo Life Sciences Supplier No. ADI901051
Add to Cart
Add to Cart
The 15(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 405 nm. Alternative name: 15(S)-hydroperoxy tetraenoic eicosatetraenoic acid. Assay Time: Overnight + 1 hour. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .