Learn More
Cayman Chemical 15(S)-Latanoprost, Purity: ≥98%, CAS: 145773-22-4, 50 mg
SDP
Catalog No. 502967141
Encompass_Preferred
A 15(S) isomer of latanoprost; reduces IOP in cynomolgus monkeys and does not induce eye irritation in cats when applied topically at 3 µg/eye; a potential impurity in commercial preparations of latanoprost
Catalog No. 50-296-7141 Supplier Cayman Chemical Supplier No. 1681850 MG
A 15(S) isomer of latanoprost; reduces IOP in cynomolgus monkeys and does not induce eye irritation in cats when applied topically at 3 µg/eye; a potential impurity in commercial preparations of latanoprost