Learn More
Chemglass Life Sciences 15 x 45 Vials, Amber, 4mL (1 Dram) Capacity, 13-425 Thread with PTFE Lined Solid PTFE Caps, Sold in a LAB-PAC of 144pcs
SDP
Catalog No. 50873522
Encompass_Preferred
15 x 45 Vials, Amber, 4mL (1 Dram) Capacity, 13-425 Thread with Solid PTFE Caps, Sold In A LAB-PAC of 144 pcs. Amber sample vials packaged in convenient LAB-PACS of 144 pieces. Made from Type I borosilicated glass. Full height partitions minimize breakage. Solid PTFE lined caps. Open top caps with septa are available under part number CG-4910.
Catalog No. 50-873-522 Supplier Chemglass Life Sciences Supplier No. CG490501
15 x 45 Vials, Amber, 4mL (1 Dram) Capacity, 13-425 Thread with Solid PTFE Caps, Sold In A LAB-PAC of 144 pcs. Amber sample vials packaged in convenient LAB-PACS of 144 pieces. Made from Type I borosilicated glass. Full height partitions minimize breakage. Solid PTFE lined caps. Open top caps with septa are available under part number CG-4910.