Learn More
Chemglass Life Sciences PTFE Blade, 15mm x 46mm
SDP
Supplier:  Chemglass Life Sciences CG206412
Encompass_Preferred
PTFE Blade, 15mm x 46mm. The 15 x 46mm blade can be used in flasks having a capacity of 100mL and above. Pictured is a fully assembled semi-micro stirrer. This item is the PTFE blade only.
Catalog No. 50-153-1426