Learn More
Perkin Elmer US LLC 150 mm, 4.6 mm I.D., 5 Quasar C18 LC Column
Catalog No. 502112969
Encompass_Preferred
Quasar C18 LC columns, based on ultra high purity silica and manufactured to tight specification, provides high efficiency separations and excellent peak shape for a wide range of compounds. Our methodology to yield high surface coverage enhances the pH stability and extends the working range. If data integrity and high productivity are top priorities for your lab, then this column can support your needs for any application.
Catalog No. 50-211-2969 Supplier Perkin Elmer US LLC Supplier No. N9308802
Quasar C18 LC columns, based on ultra high purity silica and manufactured to tight specification, provides high efficiency separations and excellent peak shape for a wide range of compounds. Our methodology to yield high surface coverage enhances the pH stability and extends the working range. If data integrity and high productivity are top priorities for your lab, then this column can support your needs for any application.