Learn More
Perkin Elmer US LLC 150 mm, 4.6 mm I.D., 5 Quasar C8 LC Column
Catalog No. 502112327
Encompass_Preferred
Quasar C8 LC columns, based on ultra high purity silica and manufactured to tight specification, offer less retention when compared to the C18 phase, due to the lower hydrophobicity. They yield high efficiency separations and excellent peak shapes for a wide range of compounds.
Catalog No. 50-211-2327 Supplier Perkin Elmer US LLC Supplier No. N9308880
Quasar C8 LC columns, based on ultra high purity silica and manufactured to tight specification, offer less retention when compared to the C18 phase, due to the lower hydrophobicity. They yield high efficiency separations and excellent peak shapes for a wide range of compounds.