Learn More
Bellco Glass, Inc. 150 x 15 mm Petri Dish, Stackable, sterile, 10/pk, 100/cs
SDP
Catalog No. 502330228
Encompass_Preferred
150 x 15 mm Petri Dish, Stackable, sterile, 10/pk, 100/cs. These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange. • High clarity polystyrene• Flat, transparent surface• Stackable for easy storage and handling• Sterilized by E-Beam• Non-pyrogenic
Catalog No. 50-233-0228 Supplier Bellco Glass, Inc. Supplier No. 451400480
150 x 15 mm Petri Dish, Stackable, sterile, 10/pk, 100/cs. These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange. • High clarity polystyrene• Flat, transparent surface• Stackable for easy storage and handling• Sterilized by E-Beam• Non-pyrogenic