Learn More
Cole-Parmer Thermo Scientific Nunc 4014-12 Petri Dish, 150 x 25 mm, 72/cs
Catalog No. NC0563201 Shop All Cole-Parmer Products
Encompass
These sterile and disposable polystyrene (PS)dishes are ideal for culturing fungi, bacteria, and other microorganisms. Dishes work well in robotic dispensers due to flatness and uniform height. Models 14007-10 and -11 are square in shape. Models 14007-03 and -05 are 5-mm deeper and models 14007-04 and -14 are 10-mm deeper than standard dish depths to accommodate longer culture periods. Sterilized by gamma irradiation.
Catalog No. NC0563201 Supplier Cole-Parmer Supplier No. 1400714
These sterile and disposable polystyrene (PS)dishes are ideal for culturing fungi, bacteria, and other microorganisms. Dishes work well in robotic dispensers due to flatness and uniform height. Models 14007-10 and -11 are square in shape. Models 14007-03 and -05 are 5-mm deeper and models 14007-04 and -14 are 10-mm deeper than standard dish depths to accommodate longer culture periods. Sterilized by gamma irradiation.