Learn More
MSC Guardair Vacuum Air Gun Kit, Pneumatic, 1-1/4 in Vac Accessories, All-Metal
Catalog No. 502839766
Encompass
This vacuum air gun kit places twice the power of a standard shop vacuum in the palm of your hand. It is powered by standard shop compressed air and has no electric motors to burn out. It features an easy-to-empty, high-filtration collection bag that attaches to the rear of the vacuum via a hose cuff. The tapered vacuum inlet accepts all standard 1-1/4 in vac accessories. The unit hangs on a hook, ready for use at any time.
  • Rugged, all-metal construction
  • Contoured comfort grip
  • Convenient hanging eyelet
  • High filtration collection bag with zipper
  • 9 in crevice tool included
  • 1/4 in FNPT bottom and rear inlets
Catalog No. 50-283-9766 Supplier MSC Supplier No. 76775485
This vacuum air gun kit places twice the power of a standard shop vacuum in the palm of your hand. It is powered by standard shop compressed air and has no electric motors to burn out. It features an easy-to-empty, high-filtration collection bag that attaches to the rear of the vacuum via a hose cuff. The tapered vacuum inlet accepts all standard 1-1/4 in vac accessories. The unit hangs on a hook, ready for use at any time.
  • Rugged, all-metal construction
  • Contoured comfort grip
  • Convenient hanging eyelet
  • High filtration collection bag with zipper
  • 9 in crevice tool included
  • 1/4 in FNPT bottom and rear inlets