Learn More
Chemglass Life Sciences 15L Duran Filtering Flask
SDP
Supplier:  Chemglass Life Sciences CG156008
Encompass_Preferred
15L Duran Filtering Flask. Duran heavy wall filtering flask has a unique removable PPN hose connection. Hose connection is secured to the flask via a silicone rubber seal. Bottle shaped filter flask, uncoated with removable PPN hose connection. For replacement PPN hose connection sets see CG-1563.
Catalog No. 50-872-077