Learn More
Ace Glass, Inc. 1500mL Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.
SDP
Catalog No. 50974290
Encompass_Preferred
1500mL Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.
Catalog No. 50-974-290 Supplier Ace Glass, Inc. Supplier No. 718672
1500mL Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.