Learn More
Ace Glass, Inc. 1500mL Buchner filter funnel with porosity D (10-20 micron) ACE fritted glass disc.
SDP
Catalog No. 50974292
Encompass_Preferred
1500mL Buchner filter funnel with porosity D (10-20 micron) ACE fritted glass disc.
Catalog No. 50-974-292 Supplier Ace Glass, Inc. Supplier No. 718678
1500mL Buchner filter funnel with porosity D (10-20 micron) ACE fritted glass disc.