Learn More
MSC PORTER-CABLE Heat Gun, 1500 Watt, Variable Temperature, 1 ea
Catalog No. 501113563
Encompass
This heat gun features a dual-fan speed selector, offering both high and low settings. A variable temperature control dial allows for easy adjustment of temperatures. The integrated hands-free support stand enables the gun to sit upright for convenient use, and its lightweight design helps reduce user fatigue, making it suitable for various tasks like paint removal, bending plastic pipes, and shrinking plastic tubing.
  • Provides 1500 watts for high heat and multiple fan settings
  • Offers 2 speed selector for high and low fan speed settings
  • Features variable temperature dial for easy adjustment of temperatures
  • Integrated support stand allows the gun to sit upright for hands-free use
  • Integrated hanging hook is ideal for storage
  • Includes 6 ft. high grade cord to improve flexibility during cold use
  • Lightweight 2 lb. gun offers less fatigue when holding
Catalog No. 50-111-3563 Supplier MSC Supplier No. 54147566
You must be signed in and have a business account to purchase online.
This heat gun features a dual-fan speed selector, offering both high and low settings. A variable temperature control dial allows for easy adjustment of temperatures. The integrated hands-free support stand enables the gun to sit upright for convenient use, and its lightweight design helps reduce user fatigue, making it suitable for various tasks like paint removal, bending plastic pipes, and shrinking plastic tubing.
  • Provides 1500 watts for high heat and multiple fan settings
  • Offers 2 speed selector for high and low fan speed settings
  • Features variable temperature dial for easy adjustment of temperatures
  • Integrated support stand allows the gun to sit upright for hands-free use
  • Integrated hanging hook is ideal for storage
  • Includes 6 ft. high grade cord to improve flexibility during cold use
  • Lightweight 2 lb. gun offers less fatigue when holding