Learn More
Sino Biological Recombinant Human T Plastin/PLS3 Protein (His Tag)
Supplier:  Sino Biological 15042H07E100
Encompass_Preferred
A DNA sequence encoding the human PLS3 (P13797) (Gly102-Asn375) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9376