Learn More
Sino Biological Recombinant Human PFDN1 Protein (His Tag)
Supplier:  Sino Biological 15092H07E100
Encompass_Preferred
A DNA sequence encoding the human PFDN1 (NP_002613.2) (Met1-Gln122) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9393