Learn More
Sino Biological Recombinant Human eIF5A Protein (His Tag)
Supplier:  Sino Biological 15096H07E100
Encompass_Preferred
A DNA sequence encoding the human EIF5A (NP_001961.1) (Met1-Lys154) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9397