Learn More
Chemglass Life Sciences 150mL Beaker, Low Form, Griffin, Pyrex
SDP
Catalog No. 501215002
Encompass_Preferred
150mL Beaker, Low Form, Griffin, Pyrex. Manufactured with uniform wall thickness these beakers offer the optimum balance between thermal shock resistance and mechanical strength. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements.
Catalog No. 50-121-5002 Supplier Chemglass Life Sciences Supplier No. CG8048P150
150mL Beaker, Low Form, Griffin, Pyrex. Manufactured with uniform wall thickness these beakers offer the optimum balance between thermal shock resistance and mechanical strength. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements.