Learn More
Ace Glass, Inc. 150mL filter reactor body with 60mm ID grooved top flange, #50 bottom thread, 60mm IDx75mm depth, with CAPFE O-ring, pressure rated at 35 psi.
SDP
Catalog No. 50972787
Encompass
Catalog No. 50-972-787 Supplier Ace Glass, Inc. Supplier No. 638405