Learn More
Chemglass Life Sciences 150mL Filter Funnel, Buchner, 29/42 Lower Vacuum Assembly, Coarse Frit
SDP
Catalog No. CG140621
Encompass_Preferred
150mL Filter Funnel, Buchner, 29/42 Lower Vacuum Assembly, Coarse Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.
Catalog No. CG140621 Supplier Chemglass Life Sciences Supplier No. CG140621
Add to Cart
Add to Cart
150mL Filter Funnel, Buchner, 29/42 Lower Vacuum Assembly, Coarse Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.