Learn More
Integra Biosciences Corp 150 ml, 100 Reservoirs (4 Sleeves of 25), Bulk, Sterile, Polypropylene
SDP
Catalog No. NC1705840
Encompass
150 ml and 300 ml Clear Advantage reagent reservoirs which are made of both crystal clear virgin polystyrene and polypropylene. Both options fit into a reusable automation friendly base with bold, crisp, clearly visible graduation markings.
This item is not returnable. View return policy
Catalog No. NC1705840 Supplier Integra Biosciences Corp Supplier No. 6338
150 ml and 300 ml Clear Advantage reagent reservoirs which are made of both crystal clear virgin polystyrene and polypropylene. Both options fit into a reusable automation friendly base with bold, crisp, clearly visible graduation markings.