Learn More
Chemglass Life Sciences 150mL Freeze Dry Flask, Crosses to Virtis P/N 148957
SDP
Catalog No. CG161122
Encompass_Preferred
150mL Freeze Dry Flask, Virtis Style, P/N 148957. Flat bottom borosilicate flask for use with and Virtis freeze dryers. Wide mouth design provides easy access sample.
Catalog No. CG161122 Supplier Chemglass Life Sciences Supplier No. CG161122
150mL Freeze Dry Flask, Virtis Style, P/N 148957. Flat bottom borosilicate flask for use with and Virtis freeze dryers. Wide mouth design provides easy access sample.