Learn More
Chemglass Life Sciences 150mL Funnel Only, Coarse Frit, #30 O-Ring Joint, 50 O.D., 50 Pinchclamp
SDP
Catalog No. CG140510
Encompass_Preferred
150mL Funnel Only, Coarse Frit, #30 O-Ring Joint, 50 O.D., 50 Pinchclamp. For positive pressure filtration of materials up to 15 psi. Connection between the funnel and adapter is made by an o-ring joint which permits a larger, grease free opening for sample retrieval. The 24/40 lower inner standard taper joint has an outlet for connection to a vacuum source.
Catalog No. CG140510 Supplier Chemglass Life Sciences Supplier No. CG140510
150mL Funnel Only, Coarse Frit, #30 O-Ring Joint, 50 O.D., 50 Pinchclamp. For positive pressure filtration of materials up to 15 psi. Connection between the funnel and adapter is made by an o-ring joint which permits a larger, grease free opening for sample retrieval. The 24/40 lower inner standard taper joint has an outlet for connection to a vacuum source.