Learn More
Chemglass Life Sciences Sublimation Apparatus, 150mL, Complete
SDP
Catalog No. 50873425
Encompass_Preferred
Sublimation Apparatus, 150mL, Complete. Two-piece grease-free sublimer with a flat bottom to enable heating on a hot plate stirrer. Condenser has a large top opening for use with dry ice / acetone or liquid nitrogen. Connection between the flask and condenser is made with a flat o-ring joint permitting parts to be gently separated for maximum product recovery. The capacity listed is that below the bottom of the cold finger. Vacuum control is provided by a 0-8mm grease-free Chem-Cap valve. Complete apparatus consists of the condenser, flask, Viton o-ring and horseshoe clamp.
Catalog No. 50-873-425 Supplier Chemglass Life Sciences Supplier No. CG303801
Sublimation Apparatus, 150mL, Complete. Two-piece grease-free sublimer with a flat bottom to enable heating on a hot plate stirrer. Condenser has a large top opening for use with dry ice / acetone or liquid nitrogen. Connection between the flask and condenser is made with a flat o-ring joint permitting parts to be gently separated for maximum product recovery. The capacity listed is that below the bottom of the cold finger. Vacuum control is provided by a 0-8mm grease-free Chem-Cap valve. Complete apparatus consists of the condenser, flask, Viton o-ring and horseshoe clamp.