Learn More
Ace Glass, Inc. 150mm powder funnel with 29/42 inner joint
SDP
Catalog No. 50974711
Encompass_Preferred
150mm powder funnel with 29/42 inner joint is useful in pouring powders or liquids into ground joint containers.
Catalog No. 50-974-711 Supplier Ace Glass, Inc. Supplier No. 723618
150mm powder funnel with 29/42 inner joint is useful in pouring powders or liquids into ground joint containers.