Learn More
Ace Glass, Inc. 4L 150mm Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.
SDP
Catalog No. 50974297
Encompass_Preferred
4L 150mm Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.
Catalog No. 50-974-297 Supplier Ace Glass, Inc. Supplier No. 7186110
4L 150mm Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.