Learn More
Ace Glass, Inc. 4L 150mm Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.
SDP
Catalog No. 50979172
Encompass_Preferred
4L 150mm Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.
Catalog No. 50-979-172 Supplier Ace Glass, Inc. Supplier No. 7186112
4L 150mm Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.