Learn More
Ace Glass, Inc. 6L 150mm Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.
SDP
Catalog No. 50974300
Encompass_Preferred
6L 150mm Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.
Catalog No. 50-974-300 Supplier Ace Glass, Inc. Supplier No. 7186130
6L 150mm Buchner filter funnel with porosity A (145-174 micron) ACE fritted glass disc.