Learn More
Ace Glass, Inc. 6L 150mm Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.
SDP
Catalog No. 50979173
Encompass_Preferred
6L 150mm Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.
Catalog No. 50-979-173 Supplier Ace Glass, Inc. Supplier No. 7186132
Add to Cart
Add to Cart
6L 150mm Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.