Learn More
Ace Glass, Inc. 6L 150mm Buchner filter funnel with porosity D (10-20 micron) ACE fritted glass disc.
SDP
Catalog No. 50974301
Encompass_Preferred
6L 150mm Buchner filter funnel with porosity D (10-20 micron) ACE fritted glass disc.
Catalog No. 50-974-301 Supplier Ace Glass, Inc. Supplier No. 7186136
6L 150mm Buchner filter funnel with porosity D (10-20 micron) ACE fritted glass disc.