Learn More
Chemglass Life Sciences Flange, Compatible with Buchi Rotary Evaporators, Flange OD: 150mm, Flange ID: 119mm, Shank OD: 130mm
SDP
Catalog No. CG14605
Encompass_Preferred
Replacement flange compatible with Buchi condensers and rotary evaporator flasks. Flange OD: 150mm, Flange ID: 119mm, Shank OD: 130mm.
Catalog No. CG14605 Supplier Chemglass Life Sciences Supplier No. CG14605
Replacement flange compatible with Buchi condensers and rotary evaporator flasks. Flange OD: 150mm, Flange ID: 119mm, Shank OD: 130mm.