Learn More
Bellco Glass, Inc. 150mm Cell Culture Dish, TC, sterile 5/pk,100/cs
SDP
Catalog No. 502330642
Encompass_Preferred
150mm Cell Culture Dish, TC, sterile 5/pk,100/cs. cell culture dishes are ideal for all types of culturing where microscopic examination is required. Numeric indicators on the bottom of eachplate allow users to identify the location of cells. • High clarity, 100% virgin polystyrene.• Flat transparent surface for distortion-free observation.• Vacuum plasma TC treatment, excellent cell adherence.• Stackable for easy storage and handling.• Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.
Catalog No. 50-233-0642 Supplier Bellco Glass, Inc. Supplier No. 451400057
150mm Cell Culture Dish, TC, sterile 5/pk,100/cs. cell culture dishes are ideal for all types of culturing where microscopic examination is required. Numeric indicators on the bottom of eachplate allow users to identify the location of cells. • High clarity, 100% virgin polystyrene.• Flat transparent surface for distortion-free observation.• Vacuum plasma TC treatment, excellent cell adherence.• Stackable for easy storage and handling.• Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.