Learn More
Chemglass Life Sciences O-Ring, FEP-Stainless Steel, 150mm Flange
SDP
Catalog No. CG14777
Encompass_Preferred
O-Ring, FEP-Stainless Steel, 150mm Flange. O-ring for Chemglass reaction vessels and lids. Available in Viton, Silicone, Perfluoro, FEP encapsulated Silicone and FEP encapsulated Stainless Steel spring. The Stainless Steel spring maintains resilience even in long term exposure to cryogenic temperatures.Temperature Range:Viton: -30 to 200°CSilicone: -50 to 230°CPerfluoro: -7 to 230°CFEP Encapsulated Silicone: -62 to 205°CFEP Encapsulated Stainless: -250 to 205°C
Catalog No. CG14777 Supplier Chemglass Life Sciences Supplier No. CG14777
O-Ring, FEP-Stainless Steel, 150mm Flange. O-ring for Chemglass reaction vessels and lids. Available in Viton, Silicone, Perfluoro, FEP encapsulated Silicone and FEP encapsulated Stainless Steel spring. The Stainless Steel spring maintains resilience even in long term exposure to cryogenic temperatures.Temperature Range:Viton: -30 to 200°CSilicone: -50 to 230°CPerfluoro: -7 to 230°CFEP Encapsulated Silicone: -62 to 205°CFEP Encapsulated Stainless: -250 to 205°C