Learn More
Chemglass Life Sciences Reaction Kettle Flange, Duran, Flat Ground, Flange ID: 150mm
SDP
Catalog No. CG14804
Encompass_Preferred
Reaction Kettle Flange, Duran, Flat Ground, Flange ID: 150mm. Similar to CG-147, but formed without the o-ring groove.
Catalog No. CG14804 Supplier Chemglass Life Sciences Supplier No. CG14804
Reaction Kettle Flange, Duran, Flat Ground, Flange ID: 150mm. Similar to CG-147, but formed without the o-ring groove.