Learn More
Chemglass Life Sciences Reaction Vessel Lid, 5-Neck, 150mm Schott Flange
SDP
Supplier:  Chemglass Life Sciences CG194640
Encompass_Preferred
Reaction Vessel Lid, 5-Neck, 150mm Schott Flange. Five neck reaction vessel lid for 150mm flanged vessels. Lid has a 29/42 outer CN, (2) 34/45 outer SN’s, (1) Vertical 45/50 outer SN and (1) 45/50 outer SN angled 20° for powder addition.
Catalog No. CG194640