Learn More
Tritech Research Inc 150MM STACK PETRI DISHES 120/C
SDP
Catalog No. NC1621945
Encompass
Extra-Sturdy Petri Dish, 150mm, Sterile, Polystyrene, Triple-vented to allow airflow, Stackable, Sturdy, Super-flat, Crystal clear
Catalog No. NC1621945 Supplier Tritech Research Inc Supplier No. T3325
Extra-Sturdy Petri Dish, 150mm, Sterile, Polystyrene, Triple-vented to allow airflow, Stackable, Sturdy, Super-flat, Crystal clear