Learn More
Bellco Glass, Inc. 150mm x 20mm Tissue Culture Treated Dish, Sterile
SDP
Catalog No. 502329900
Encompass_Preferred
Cell Culture Dishes have thicker and more durable plastic than Petri DishesVented lids contain molded spacers to allow gas exchange Pack Description5/Re-sealable BagCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume25-28 mLTissue Culture TreatedYesCell Growth Area143cm²RNase/DNase FreeYes
Catalog No. 50-232-9900 Supplier Bellco Glass, Inc. Supplier No. 229651000
Cell Culture Dishes have thicker and more durable plastic than Petri DishesVented lids contain molded spacers to allow gas exchange Pack Description5/Re-sealable BagCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneWorking Volume25-28 mLTissue Culture TreatedYesCell Growth Area143cm²RNase/DNase FreeYes