Learn More
CELLTREAT Scientific Products 150mm x 25mm Tissue Culture Treated Dish, Sterile, 60/CS
SDP
Catalog No. 501535084
Encompass_Preferred
CELLTREAT Scientific Products 150mm x 25mm Tissue Culture Treated Dish. Tall dish allows for the use of more media or provides extra headspace. Cell culture treated hydrophilic surface for enhanced cell attachment. Vented lid contains molded spacers to allow gas exchange. Manufactured from virgin polystyrene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/bag, 60/case.
Catalog No. 50-153-5084 Supplier CELLTREAT Scientific Products Supplier No. 229652
CELLTREAT Scientific Products 150mm x 25mm Tissue Culture Treated Dish. Tall dish allows for the use of more media or provides extra headspace. Cell culture treated hydrophilic surface for enhanced cell attachment. Vented lid contains molded spacers to allow gas exchange. Manufactured from virgin polystyrene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/bag, 60/case.