Learn More
CELLTREAT Scientific Products 150mm x 15mm Petri Dish, Sterile
SDP
Catalog No. 501149058
Encompass_Preferred
CELLTREAT Scientific Products 150mm x 15mm Non-Treated Petri Dish. Dishes have thicker and more durable plastic than other Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 25-28mL and growth area of 153cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/re-sealable bag, 100/case.
Catalog No. 50-114-9058 Supplier CELLTREAT Scientific Products Supplier No. 229656
CELLTREAT Scientific Products 150mm x 15mm Non-Treated Petri Dish. Dishes have thicker and more durable plastic than other Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 25-28mL and growth area of 153cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/re-sealable bag, 100/case.